Learn More
Norgen Biotek Corp Total RNA Purification Micro Kit, 50 Prep
Supplier:  Norgen Biotek Corp 35300
Encompass_Preferred
This kit extracts total RNA from low cell number samples and elutes in a convenient 20 µL elution for a number of downstream applications.This kit is suitable for the isolation of total RNA from a range of samples including small input of cells, needle biopsies, LCM, CTC and other low cell number samples. Extract high quality and purity RNA with excellent RIN values and A260/A280 suitable for downstream applications including qRT-PCR, RT-PCR, microarrays, NGS and more. The kit purifies all sizes of RNA from large mRNA, lncRNA down to microRNA (miRNA) in the same fraction without the requirement of phenol. Isolate all RNA sequences at an equal rate irrespective of size. Moreover, when the RNA sequences are small (e.g. miRNA), the column binds small RNAs regardless of their GC content.
Catalog No. NC0623526