Learn More
MICROGENICS CORPORATION DRI Total Thyroxine Assay, 500 mL kit, quantitative determination of total thyroxine in human serum or plasma, ready-to-use liquid reagents, CLIA Non-Waived
Catalog No. NC2676148
Encompass
The Microgenics DRI Total Thyroxine (T4) Assay is a homogeneous enzyme immunoassay designed for the quantitative determination of total thyroxine in human serum or plasma. This assay streamlines laboratory workflows with its ready-to-use liquid reagents and boasts a high correlation with other T4 assays. It is classified as CLIA Non-Waived, ensuring it meets the rigorous standards for more complex tests.
  • Quantitative determination of total thyroxine
  • Homogeneous enzyme immunoassay
  • Ready-to-use liquid reagents
  • Strong correlation coefficient of 0.993 with other T4 assays
  • Compatible with clinical chemistry analyzers
  • Requires refrigeration for storage
  • Sensitivity of 0.7 μg/dL
  • In vitro diagnostic use
  • Sample types include human serum and plasma
This item is not returnable. View return policy
Catalog No. NC2676148 Supplier MICROGENICS CORPORATION Supplier No. 10013070
The Microgenics DRI Total Thyroxine (T4) Assay is a homogeneous enzyme immunoassay designed for the quantitative determination of total thyroxine in human serum or plasma. This assay streamlines laboratory workflows with its ready-to-use liquid reagents and boasts a high correlation with other T4 assays. It is classified as CLIA Non-Waived, ensuring it meets the rigorous standards for more complex tests.
  • Quantitative determination of total thyroxine
  • Homogeneous enzyme immunoassay
  • Ready-to-use liquid reagents
  • Strong correlation coefficient of 0.993 with other T4 assays
  • Compatible with clinical chemistry analyzers
  • Requires refrigeration for storage
  • Sensitivity of 0.7 μg/dL
  • In vitro diagnostic use
  • Sample types include human serum and plasma