Learn More
Crystal Chem Inc Total Thyroxine (T4) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80983
Encompass_Preferred
The Total Thyroxine ELISA is a highly sensitive assay that is used to quantify levels of total human thyroxine in serum. Highlights: Kits use sample size of only 20 µL. High sensitivity of 30 5 µIU/mL. Dynamic range of 0.2 - 30 µIU/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7884