Learn More
Biolegend TotalSeq™-C Mouse Universal Cocktail, V1.0, Lyophilized, 5 tests, Mouse reactivity, TotalSeq™-C conjugate, Primary antibody, 10x Genomics Single Cell 5' workflow compatible, Flex Gene Expression compatible, Cell staining buffer recommended for reconstitution
Supplier:  Biolegend 199903
Encompass_Preferred
TotalSeq™-C Mouse Universal Cocktail, V1.0, Lyophilized, 5 tests, Mouse reactivity, TotalSeq™-C conjugate, Primary antibody, 10x Genomics Single Cell 5' workflow compatible, Flex Gene Expression compatible, Cell staining buffer recommended for reconstitution
Catalog No. NC2213788
Only null left
Add to Cart
Request a Quote