Learn More
Biolegend TotalSeq™-C0088 anti-human CD279 (PD-1)
Supplier:  Biolegend 329963
Encompass_Preferred
TotalSeq™-C0088 anti-human CD279 (PD-1) [EH12.2H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Squirrel Monkey; Apps: PG; Size: 10 ug
Catalog No. NC1624623