Learn More
Biolegend TotalSeq™-C0301 anti-mouse Hashtag 1
Supplier:  Biolegend 155861
Encompass_Preferred
TotalSeq™-C0301 anti-mouse Hashtag 1 [M1/42; 30-F11]; Isotype: Rat IgG2a, κ/Rat IgG2b, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. NC1920796