Learn More
Biolegend TotalSeq™-C0306 anti-mouse Hashtag 6
Supplier:  Biolegend 155871
Encompass_Preferred
TotalSeq™-C0306 anti-mouse Hashtag 6 [M1/42; 30-F11]; Isotype: Rat IgG2a, κ/Rat IgG2b, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. NC1920788