Learn More
Biolegend TotalSeq™-C0831 anti-human CD138 (Syndecan-1)
Supplier:  Biolegend 352327
Encompass_Preferred
TotalSeq™-C0831 anti-human CD138 (Syndecan-1) [DL-101]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: PG; Size: 10 ug
Catalog No. 50-207-7131