Learn More
Biolegend TotalSeqTM-Bn1337 anti-Granzym
Supplier:  Biolegend 674605
Encompass_Preferred
TotalSeq™-Bn1337 anti-Granzyme B [M3304B06]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: SB; Size: 10 μg
Catalog No. 50-274-5653