Learn More
Biolegend TotalSeqTM-C0926 anti-mouse CD
Supplier:  Biolegend 151127
Encompass_Preferred
TotalSeq™-C0926 anti-mouse CD186 (CXCR6) [SA051D1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: PG; Size: 10 μg
Catalog No. 50-272-2620