Learn More
Biolegend TotalSeq™-C1373 anti-human CD97 [VIM3b]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 μg
Supplier:  Biolegend 336317
Encompass_Preferred
TotalSeq™-C1373 anti-human CD97 [VIM3b]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 μg
Catalog No. 50-272-3309