Learn More
MSC Rochester Midland Corporation Cleaner & Degreaser, Liquid, Spray, 1 qt, Mild Scent
Supplier:  MSC 36099372
Encompass
The Rochester Midland Corporation Cleaner & Degreaser is a versatile, ready-to-use solution designed for effective cleaning across various surfaces. It is a water-based formula with a mild scent, suitable for industrial and commercial applications.
  • Formulated as a liquid and spray for versatile application.
  • Ready to use, no dilution required.
  • Mild scent for user comfort.
  • Water based composition.
  • Removes a wide range of contaminants including grease, oil, dirt, and carbon residue.
  • Suitable for cleaning walls, machinery, parts, and all hard surfaces.
  • EcoLogo and USDA Certified BioBased standards met.
Catalog No. 50-283-6448