Learn More
Research Products International Corp Tough-Spots for Laser Printers, 1/2 Inch Diameter, White, 3,840 per Package
SDP
Supplier:  Research Products International Corp 247139
Encompass
Tough-Spots are chemically resistant labels that adhere to polypropylene tubes without cracking, peeling or degrading. Specially designed to withstand boiling water baths, autoclaving freezing, cryogenic liquid nitrogen storage, organic solvents and caustic agents. Tough-Spots are made of a special smear resistant material and will accept any marking instrument.
Catalog No. 50-212-524