Learn More
Medchemexpress LLC Tovinontrine | 2062661-53-2 | 99.8% | 394.47 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY109193100MG
Encompass_Preferred
Tovinontrine (IMR-687) is described as a highly potent and selective phosphodiesterase-9 (PDE9) inhibitor. It is specifically intended for the treatment of sickle cell disease. The compound exhibits IC50 values of 8.19 nM for PDE9A1 and 9.99 nM for PDE9A2. It is for research use only and not sold to patients.
  • Highly potent and selective PDE9 inhibitor
  • Potential treatment for sickle cell disease
  • Induces hemoglobin (HbF)
  • Increases cGMP levels
  • Reduces sickled red blood cells and hemolysis in vivo
Catalog No. 50-003-45424