Learn More
Genscript Corporation ToxinEraser™ Endotoxin Removal Kit
Supplier:  Genscript Corporation L00338
Encompass
GenScript ToxinEraser™ is an endototoxin removal resin of high efficiency. It is based on the affinity matrix of modified polymyxin B (PMB), which allows highly efficient endotoxin removal. The final endotoxin level can be decreased to less than 0.1 EU/ml with repeat use of ToxinEraser™ endotoxin removal resin. Final removal efficiency may vary depending on the sample type/source.
Catalog No. NC0853916