Learn More
Cayman Chemical ToyocamycInhydrate 5mg
SDP
Supplier:  Cayman Chemical 173715 MG
Encompass_Preferred
A natural adenosine analog that prevents IRE1α-induced mRNA cleavage (IC50 = 80 nM) and inhibits constitutive activation of XBP1 in multiple myeloma cell lines; used to study IRE1α action in the endoplasmic reticulum stress response
Catalog No. 50-296-5845