Learn More
Tosoh Bioscience LLC TOYOPEARL HEXYL-650C 25ML
Supplier:  Tosoh Bioscience LLC 0044465
Encompass
Media, Bulk Resins; For Hydrophobic Interaction Chromatography; Tosoh Toyopearl Hexyl-650C, 100 micrometer; Matrix: Polymer; Container Size: 25mL
Catalog No. 50-851-753