Learn More
Tosoh Bioscience LLC TOYOPEARL HW-75S 35UM 250ML
Supplier:  Tosoh Bioscience LLC 0007471
Encompass
Media, Bulk Resins; For Size Exclusion Chromatography; Tosoh Toyopearl HW-75S, 30 micrometer; Matrix: Polymer; Container Size: 250mL
Catalog No. 50-851-201