Learn More
Tosoh Bioscience LLC TOYOPEARL PHENYL-650S 100ML
Supplier:  Tosoh Bioscience LLC 0014477
Encompass
Media, Process Resins; For Hydrophobic Interaction Chromatography; Tosoh Toyopearl Phenyl-650S, 35 micrometer; Matrix: Polymer; Container Size: 100mL
Catalog No. 50-851-368