Learn More
Medchemexpress LLC TP-064 | 10MG
SDP
Supplier:  Medchemexpress LLC HY11496510MG
Encompass_Preferred
TP-064 a chemical probe is a potent and selective proteinarginine methyltransferase 4 (PRMT4 CARM1) inhibitor (IC50 10 nM) TP-064 inhibits dimethylation of BAF155 (IC50 of 340 nM) and MED12 (IC50 of 43 nM) TP-064 is inactive against the other family members except for PRMT6 (IC50 of 1 3 M) TP-064 has anticancer activities[1]
Catalog No. 50-003-07563