Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TP-064 50mg | 2080306-20-1 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY11496550MG

Encompass_Preferred

TP-064 is a potent, selective small-molecule inhibitor of protein arginine methyltransferase 4 (PRMT4/CARM1) supplied for research use. It inhibits PRMT4 with high potency, affects dimethylation of substrates such as BAF155 and MED12, and is provided as a high-purity powder with recommended cold-storage conditions.

  • Potent PRMT4 inhibitor (IC50 <10 nM).
  • Modulates dimethylation of BAF155 and MED12.
  • High purity (~99.6%).
  • Molecular formula C28H34N4O2; molecular weight 458.6 g/mol.
  • Available in small pack sizes suitable for biochemical and cellular studies.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations.

Catalog No. 50-002-28930


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.