Learn More
Abcam TP-3654, 25MG
Supplier:  Abcam AB28695925MG
Encompass_Preferred
MW 418.5 Da, Purity >=98%. A potent second-generation Pim kinase inhibitor with Ki values of 5 and 42 nM for Pim-1 and Pim-3, respectively. Displays anti-cancer activities.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5799