Learn More
TARGETMOL CHEMICALS INC TPCA-1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T304925MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. TPCA-1 (TPCA1) is an effective and specific IKK-2 inhibitor (IC50 17.9 nM); shows >22-fold selectivity over IKK-1 and >550-fold selectivity over others kinases and enzymes. purity: 99%
Catalog No. 50-258-0413