Learn More
TARGETMOL CHEMICALS INC TPCK 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T8350200MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TPCK is an irreversible inhibitor of chymotrypsin-like proteases that affect cell proliferation, apoptosis, and tumorigenesis Purity 96.77%
Catalog No. 50-287-9161