Learn More
Cayman Chemical TPEN, 16858-02-9, 10 mg
SDP
Supplier:  Cayman Chemical 1334010
Encompass_Preferred
Molecular Formula C26H28N6, Purity ≥95%, Formula Weight 424.6, Synonyms: TPEDA; A metal ion chelator used in zinc treatment studies to limit intra- and extracellular concentrations of the minerial.
Catalog No. NC0388288