Learn More
Med Vet International TPLO Locking Plate 3.5 Broad Pre-contoured Right, J0438ALD
Supplier:  Med Vet International JOR438ALD
Encompass_Preferred
The three screws in the head of the standard TPLO locking plate are concentrated into a relatively small area. The screws of the locking TPLO plate are more widely spread out along the osteotomy offering better support of the proximal tibial segment. The two outlying screws converge and the central screw angles into a different plane for maximum re- sistance. The osteotomy based plate means that the osteotomy may be place high relative to standard locking TPLO plates. Available in 3.5 broad, 3.5 narrow and 2.7mm. Plates are pre-contoured.3.5mm Broad Locking TPLO Right (left leg)
Catalog No. 50-005-82663