Learn More
TARGETMOL CHEMICALS INC TPN171 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T131935MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TPN171 is potent PDE5 inhibitor with subnanomolar potency for PDE5 and good selectivity over PDE6 which has the potential for the treatment of pulmonary arterial hypertension (PAH). TPN171 was proven to exert a longer lasting effect than sildenafil in animal models providing a foundation for a once-daily oral administration for its clinical use. purity: 100%
Catalog No. 50-258-1370