Learn More
Medchemexpress LLC TPN729MA 10mM 1mL | 1422955-52-9 | 632.73 g/mol | C29H40N6O8S | 1 ML

Supplier: Medchemexpress LLC HY116808B10MM 1ML
TPN729MA (TPN729 maleate) is a selective phosphodiesterase type 5 (PDE5) inhibitor supplied for research use. It is provided as a maleate salt and is available as a 10 mM stock solution in DMSO (1 mL) and in multiple solid sizes.
- Selective phosphodiesterase type 5 (PDE5) inhibitor.
- Provided as a maleate salt for improved handling.
- Available as a 10 mM stock solution in DMSO, 1 mL.
- Also available in solid quantities (5 mg-100 mg).
- Molecular formula: C29H40N6O8S.
- Molecular weight: 632.73 g/mol.
- Purity: 99.5% (supplier stated).
- Storage: solid at 4°C; in solvent, -80°C up to 6 months or -20°C up to 1 month.
- CAS number: 1422955-52-9.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.