Learn More
Medchemexpress LLC TPN729MA 5mg | 1422955-52-9 | 632.73 | C29H40N6O8S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY116808B5MG
Encompass_Preferred
TPN729MA (TPN729 maleate) is a selective phosphodiesterase type 5 (PDE5) inhibitor supplied as the maleate salt for laboratory research use only. It is provided at high purity and is intended for biochemical and cellular studies of PDE5 activity and erectile dysfunction-related signaling.
  • Selective PDE5 inhibitor with reported IC50 = 2.28 nM.
  • Supplied as the maleate salt for improved handling and solubility.
  • High purity (99.45%) suitable for research assays.
  • Available as small-mass solid vials and DMSO stock solutions.
  • Intended for in vitro research use only, not for human consumption.
Catalog No. 50-002-28280