Learn More
Medchemexpress LLC TPO/Thyroid Peroxida 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7804550UG
Encompass_Preferred
Thyroid Peroxidase (TPO) orchestrates iodination and coupling within thyroglobulin leading to the synthesis of thyroid hormones T(3) and T(4) TPO/Thyroid Peroxidase Protein Mouse (HEK293 His) is the recombinant mouse-derived TPO/Thyroid Peroxidase protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87808