Learn More
Cole-Parmer TPE Tubing, Black, 1/4" ID x 1/2" OD; 25 Ft
Supplier:  Cole-Parmer 5012127
Encompass_Preferred
Cole-Parmer TPE tubing is a high-performance industrial tubing manufactured from a thermoplastic elastomeric compound that consistently outperforms neoprene, EPDM and other general purpose rubber tubing. The Cole-Parmer TPE tubing features an extended service life in a wide range of applications including caustic chemical transfer, wastewater sampling, soap and disinfectant dispensing, vacuum pumps, glass and window washing systems and more. This tubing is opaque black, allowing for light and ozone resistance without weakening or cracking, and can withstand extreme temperatures from -75°F to 275°F (-60 to 135°C).
Catalog No. 50-255-7707