Learn More
Hach Company TPTZ Iron Reagent Powder Pillows, 10 mL, pk/100
Supplier:  Hach Company 2608799
Encompass
For low range Iron determination by the TPTZ method. Hach Method 8112. Range: 0.012 - 1.800 mg/L Fe. Sample Size: 10 mL. Pack of 100 Powder Pillows
Catalog No. 50-199-6338