Learn More
Cayman Chemical TPU-0037A 2.5mg
SDP
Supplier:  Cayman Chemical 257612.5 MG
Encompass_Preferred
An antibiotic; inhibits the growth of Gram-positive bacteria, including MRSA, B. subtilis, and M. luteus (MICs = 1.56-12.5 μg/ml)
Catalog No. 50-293-6057