Learn More
STEMCELL Technologies 3,3',5-Triiodo-L-thyronine (Sodium Salt Hydrate), Size: 100 mg
SDP
Supplier:  STEMCELL Technologies 1000548
Encompass
3,3',5-Triiodo-L-thyronine is a thyroid hormone, which is converted from the prohormone thyroxine via deiodination (Misiti et al.). 3,3',5-Triiodo-L-thyronine binds thyroid hormone receptors TRα and TRβ (Kd = 0.06 nM for both thyroid hormone receptors; Sandler et al.), and is essential for growth and differentiation of a variety of cell types (Misiti et al.; Shiohara et al.). 3,3',5-Triiodo-L-thyronine inhibits leucine transport by pituitary cells (IC50 = 2 μM; Yan & Hinkle). This product is supplied as the sodium salt hydrate form of the molecule.DIFFERENTIATION· Promotes pancreatic β cell differentiation from human pluripotent stem cells (Pagliuca et al.).CANCER RESEARCH· Inhibits the proliferation of pancreatic adenocarcinoma (Michienzi et al.).
Catalog No. 50-206-9356