Learn More
Thomas Scientific Traceabler Refrigerator / Freezer Thermometer w / Bottle Probe
SDP
Supplier:  Thomas Scientific 9327L12
Encompass
Accurately measures temperature in freezers, refrigerators, incubators, water baths and heating blocks Enclosed “temperature buffered” sensor Range: -50 to 70°C (-58 to 158°F) Resolution: 1°C; accuracy: ±1°C Glycol bottle insulates sensor from rapid temperature changes High-impact, chemical-resistant ABS case Min/Max memory monitors conditions overnight, weekends or any time period. An alarm alerts user when temperature rises above or falls below a set point. The alarm is programmable in 1° increments. The visual and audio signal continue even if temperature returns to non-alarm range. Micro cable is thin enough to allow refrigerator doors to be shut on it. Probe with solid state sensor is sealed in a miniature bottle filled with patented non-toxic glycol that is recognized as safe by the FDA. Thermometer includes Traceable certificate; Velcro™ and magnetic strips for mounting thermometer and bottle sensor; and one AA battery good for o
Catalog No. NC0525578