Learn More
Thomas Scientific Traceable Hygrometer / Thermometer
SDP
Supplier:  Thomas Scientific 1235C78
Encompass
Meter may be used with bench stand and can also be wall mounted. Unit measures 3 1/2” x 5” x 7/8” and weighs six ounces. Devices are provided with an individually serial numbered Traceable certificate from an ISO 17025 calibration laboratory accredited by A2LA. Certificate indicates traceability to standards provided by NIST.
Catalog No. NC1775105