Learn More
Research Products International Corp Traceable pH Buffer Standards, 10.012 pH Value, 16oz
SDP
Supplier:  Research Products International Corp 844282
Encompass
Traceable pH Buffers provide the most precise accuracy available, ± 0.010 pH @ 25°C. These buffers have a Certified Reference Materials Classification and meet all Federal, State and local requirements to provide exact calibration results with all pH meters. Each buffer is supplied with an individual and serialized Traceable Certificate of Calibration. Available in economical 16 ounce bottle featuring exclusive triple layer packaging that ensures a two year shelf life.
Catalog No. 50-197-8489