Learn More
Perkin Elmer US LLC Traceable Reference Material (TRM) - Mid-Infrared
Supplier:  Perkin Elmer US LLC L1365334
Encompass_Preferred
This reference material mounts in your FT-IR spectrometer, allowing you to quickly and easily check the mid infrared frequencies and overall accuracy of your FT-IR spectrometer.
Catalog No. 50-211-1648