Learn More
MSC U-Power Safety Boots, Full-Grain Leather, S3 SRC, 1 ea
Supplier: MSC 79508362
The Track L S3 SRC safety boots are designed to meet rigorous safety standards while providing comfort and protection in professional environments. These boots are constructed with a full-grain water-resistant leather upper and feature a breathable membrane in the safety toe cap.
- Full-grain water-resistant leather upper
- WingTex air tunnel textile lining material
- AirToe composite perforated safety toe cap with breathable membrane
- Classic steel midsole
- Anatomic, breathable footbed with EVA support
- Antislip, antistatic, and hard-wearing outer sole
- Certified to EN ISO 20344:2021 and compliant with Regulation (EU) 2016/425
- Available in sizes 35 to 48