Learn More
Medchemexpress LLC HY-108294 100mg Medchemexpress, Nonaethylene glycol monododecyl ether CAS:3055-99-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108294100MG
Encompass_Preferred
Medchemexpress, HY-110247 10mg TRAF-STOP inhibitor 6877002 CAS:433249-94-6 TRAF-STOP inhibitor 6877002, is a selective inhibitor of CD40-TRAF6 interaction, compound VII, shows inhibition of NF-κB activation in RAW cells, extracted from patent WO2014033122A1. TRAF-STOP 6877002 prevents the progression of established atherosclerosis in mice, reduces leukocyte recruitment and reduces macrophage activation; reduces macrophage proliferation in atherosclerotic plaques. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8519