Learn More
ABclonal Technology TRAF7 Rabbit pAb
SDP
Supplier:  ABclonal Technology A156071000UL
Encompass_Preferred
Tumor necrosis factor (TNF, see MIM 191160) receptor-associated factors, such as TRAF7, are signal transducers for members of the TNF receptor superfamily (see MIM 191190). TRAFs are composed of an N-terminal cysteine/histidine-rich region containing zinc RING and/or zinc finger motifs, a coiled-coil (leucine zipper) motif, and a homologous region that defines the TRAF family, the TRAF domain, which is involved in self-association and receptor binding.
Catalog No. 50-001-07583