Learn More
Medchemexpress LLC HY-10999 100mg , Trametinib GSK1120212;JTP-74057 CAS:871700-17-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10999100MG
Encompass_Preferred
Medchemexpress, HY-10999 100mg Trametinib GSK1120212;JTP-74057 CAS:871700-17-3 Formula:C26H23FIN5O4 IC50: 2 nM (MEK1/2) Purity:98% Trametinib is a potent MEK inhibitor that specifically inhibits MEK1/2, with an IC 50 value of about 2 nM. Due to the poor solubility of Trametinib, Trametinib DMSO solvate (Cat. No.: HY-10999A) is the more commonly used form. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2661