Learn More
Enzo Life Sciences Trametinib (1g). CAS: 871700-17-3
Supplier: Enzo Life Sciences ENZCHM2391000
Trametinib is a highly specific and potent MEK1/2 inhibitor with IC50 of 0.92 nM/1.8 nM in cell-free assays. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (20 mg/mL). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.