Learn More
Medchemexpress LLC Trametinib (DMSO solvate) | 1187431-43-1 | MFCD21609250 | 99.6% | 693.53 g/mol | C28H29FIN5O5S | 5 MG

Supplier: Medchemexpress LLC HY10999A5MG
Trametinib (DMSO solvate) is a potent, selective MEK1/2 inhibitor supplied as a DMSO solvate crystalline powder for research use. It is characterized for in vitro and in vivo applications with documented solubility and storage conditions; handle and formulate according to supplier guidance.
- Potent, selective MEK1/2 inhibition for signaling pathway studies.
- Supplied as a DMSO solvate crystalline powder for stability.
- Soluble in DMSO at approximately 3.33 mg/mL (4.80 mM); may require sonication.
- Formulations for in vivo use achievable at ≥ 2.5 mg/mL in common vehicles.
- Storage: powder stable at -20°C (3 years) or 4°C (2 years); solutions require low-temperature storage.
- Characterized by CAS 1187431-43-1, formula C28H29FIN5O5S, molecular weight 693.53.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.