Learn More
TARGETMOL CHEMICALS INC TRAMETINIB DMSO SOLVATE 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5857200MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Trametinib (DMSO solvate) (GSK-1120212 (DMSO solvate)) is a Highly Potent and Selective MEK Inhibitor that specifically inhibits MEK1/2(IC50 2 nM)purity: 99%
Catalog No. 50-257-6654