Learn More
Cayman Chemical TramIprosate 25g
SDP
Supplier:  Cayman Chemical 2070125 G
Encompass_Preferred
A sulfated glycosaminoglycan mimetic that inhibits Aβ aggreagation, yet promotes tau polymerization; analog of GABA that can act as a partial agonist of GABAA receptors and an antagonist at GABAB receptors
Catalog No. 50-298-3923