Learn More
TARGETMOL CHEMICALS INC Trandolapril 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T532925MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Trandolapril is an orally administered angiotensin converting enzyme (ACE) inhibitor that is hydrolysed to the active diacid trandolaprilat. Purity 99.7%
Catalog No. 50-287-7347