Learn More
Medchemexpress LLC Tranexamic acid impurity 1 | 1197-17-7 | MFCD19706018 | >=96.0% | 157.21 g/mol | C8H15NO2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0631911G
Encompass_Preferred
Tranexamic acid impurity 1 is a characterized impurity of tranexamic acid supplied as an analytical reference standard for research use. It is intended as a drug intermediate or analytical standard for qualitative and quantitative assays.
  • Used as an analytical reference standard and drug intermediate
  • Matches molecular formula C8H15NO2 and molecular weight 157.21 g/mol
  • CAS number 1197-17-7 for unambiguous identification
  • Store under conditions specified in the certificate of analysis
Catalog No. 50-002-10394