Learn More
Apexbio Technology LLC Tranilast 53902-12-8 100mg
SDP
Supplier:  Apexbio Technology LLC A5375100
Encompass_Preferred
Tranilast (CAS 53902-12-8) is a small molecule known to exert antiallergic effects by inhibiting the release of chemical mediators from mast cells Recent studies have demonstrated its capacity to suppress angiogenesis indicating an impact on vascular remodeling processes Additionally Tranilast has been shown to counteract angiotensin II-induced responses in human arterial tissue potentially through modulation of signaling pathways at the vascular level This pharmacological profile makes Tranilast a valuable tool for investigating mechanisms of allergic reactions neovascularization and vascular dysfunction in biomedical research
Catalog No. 50-224-0490