Learn More
Medchemexpress LLC Trans-1,2-Cyclohexanediaminetetraacetic acid | 13291-61-7 | 97.0% | C14H22N2O8 | 10 G
SDP
Supplier:  Medchemexpress LLC HY4529010G
Encompass_Preferred
Trans-1,2-Cyclohexanediaminetetraacetic acid is a commonly used aminopolycarboxylic acid and a strong chelator of heavy metal ions. This product is for research use only and has not been fully validated for medical applications.
  • Commonly used aminopolycarboxylic acid
  • Strong chelator of heavy metal ions
  • For research use only
  • Not fully validated for medical applications
Catalog No. 50-003-59059